Merck
  • Survey of captive parrot populations around Port Phillip Bay, Victoria, Australia, for psittacine beak and feather disease virus, avian polyomavirus and psittacine adenovirus.
Survey of captive parrot populations around Port Phillip Bay, Victoria, Australia, for psittacine beak and feather disease virus, avian polyomavirus and psittacine adenovirus.
Australian veterinary journal (2015-07-30)
C L Hulbert, A Chamings, K A Hewson, P A Steer, M Gosbell, A H Noormohammadi
ABSTRAKT
This study investigated the prevalence of psittacine beak and feather disease virus (BFDV), avian polyomavirus (APV) and psittacine adenovirus (PsAdV) in captive psittacine birds around Port Phillip Bay, Victoria, Australia. Samples of fresh droppings were collected from 118 psittacine birds (109 clinically normal and 9 with feather abnormalities) from 11 avaries in different locations and were used for detection of BFDV, APV and PsAdV using PCR. BFDV, APV and PsAdV were detected in 31%, 13% and 4%, respectively, of the specimens tested. One budgerigar was found to be co-infected with BFDV and PsAdV. At least one sample tested positive for BFDV at each location. This is the first report of the prevalence of BFDV, APV and PsAdV in Victoria and provides a foundation for future studies examining the influence of these viruses on the health of aviary birds in Victoria.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis