Merck
  • TNF receptor superfamily member 13b (TNFRSF13B) hemizygosity reveals transmembrane activator and CAML interactor haploinsufficiency at later stages of B-cell development.
TNF receptor superfamily member 13b (TNFRSF13B) hemizygosity reveals transmembrane activator and CAML interactor haploinsufficiency at later stages of B-cell development.
The Journal of allergy and clinical immunology (2015-06-24)
Neil Romberg, Manmeet Virdee, Nicolas Chamberlain, Tyler Oe, Jean-Nicolas Schickel, Tiffany Perkins, Tineke Cantaert, Rima Rachid, Sally Rosengren, Regina Palazzo, Raif Geha, Charlotte Cunningham-Rundles, Eric Meffre
ABSTRAKT
Heterozygous C104R or A181E TNF receptor superfamily member 13b (TNFRSF13B) mutations impair removal of autoreactive B cells, weaken B-cell activation, and convey to patients with common variable immune deficiency (CVID) an increased risk for autoimmunity. How mutant transmembrane activator and CAML interactor (TACI) influences wild-type TACI function is unclear; different models suggest either a dominant negative effect or haploinsufficiency. We investigated potential TACI haploinsufficiency by analyzing patients with antibody-deficient Smith-Magenis syndrome (SMS) who possess only 1 TNFRSF13B allele and antibody-deficient patients carrying one c.204insA TNFRSF13B null mutation. We tested the reactivity of antibodies isolated from single B cells from patients with SMS and patients with a c.204insA TNFRSF13B mutation and compared them with counterparts from patients with CVID with heterozygous C104R or A181E TNFRSF13B missense mutations. We also assessed whether loss of a TNFRSF13B allele induced haploinsufficiency in naive and memory B cells and recapitulated abnormal immunologic features typical of patients with CVID with heterozygous TNFRSF13B missense mutations. We found that loss of a TNFRSF13B allele does not affect TACI expression, activation responses, or establishment of central B-cell tolerance in naive B cells. Additionally, patients with SMS and those with a c.204insA TNFRSF13B mutation display normal regulatory T-cell function and peripheral B-cell tolerance. The lack of a TNFRSF13B allele did result in decreased TACI expression on memory B cells, resulting in impaired activation and antibody secretion. TNFRSF13B hemizygosity does not recapitulate autoimmune features of CVID-associated C104R and A181E TNFRSF13B mutations, which likely encode dominant negative products, but instead reveals selective TACI haploinsufficiency at later stages of B-cell development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)