Merck
Effects of Cydonia oblonga Miller leaf and fruit flavonoids on blood lipids and anti-oxydant potential in hyperlipidemia rats.
Journal of ethnopharmacology (2015-05-03)
Anwar Umar, Guldiyar Iskandar, Ainiwaer Aikemu, Wuliya Yiming, Wenting Zhou, Bénedicte Berké, Bernard Begaud, Nicholas Moore
ABSTRAKT
To study the effects of Cydonia oblonga Miller (COM) total flavonoids (TF) from leaves and fruit on the blood lipid and antioxidant potentials using hyperlipidaemic rat models. Hyperlipidaemic rat models were created with high-lipid emulsion. Rats were distributed into normal controls, hyperlipidaemic models, and daily high (160mg/kg), medium (80mg/kg) and low (40mg/kg) TF from leaves and fruit and simvastatin (5mg/kg) groups. After four weeks, serum total cholesterol (TC), triglyceride (TG), high-density lipoprotein cholesterol (HDL), low-density lipoprotein cholesterol (LDL), alanine aminotransferase (ALT), aspartate aminotransferase (AST), as well as hepatic superoxide dismutase (SOD), glutathione peroxidase (GSH-Px) and malondialdehyde (MDA) were measured. Compared with the hyperlipidaemic model group, TF significantly reduced serum TC, TG, LDL-C (P<0.01), ALT and AST (P<0.01 or P<0.05) and increased HDL-C (P<0.05 or P<0.01). TF also reduced MDA (P<0.01 or P<0.01). COM total flavonoids can effectively regulate the metabolism of lipids, and remove oxygen free radicals. This confirms its potential value in the prevention and treatment of hyperlipidaemia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%