Merck
Anti-hyperbilirubinemic and wound healing activity of aqueous extract of Calotropis procera leaves in Wistar rats.
Indian journal of pharmacology (2015-08-20)
Rupali Arun Patil, Aakash B Makwana
ABSTRAKT
The aim of this study was to evaluate the bilirubin lowering and wound healing property of aqueous extract of Calotropis procera (AECP) leaves in Wistar rats. Albino Wistar rats of either sex were used for the study. Bilirubin lowering property of C. procera leaves was evaluated using phenylhydrazine and paracetamol as inducing agents followed by measuring the concentration of serum total bilirubin in hyperbilirubinemic rats. Wound healing property was evaluated using incision and excision models by measuring tensile breaking strength, percentage wound contractions, and epithelization days, respectively. Statistical comparison between groups in each experiment was done with one-way analysis of variance followed by Dunnett's test. AECP showed a significant (P < 0.05) decrease in concentrations of serum total bilirubin in hyperbilirubinemic rats as well as significant (P < 0.05) increase in breaking strength and percentage wound contractions with decreased epithelization period when compared to control groups. AECP showed significant bilirubin lowering and wound healing property in Wistar rats.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phenylhydrazine, 97%
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications