Merck
The impact of anti-tuberculous antibiotics and corticosteroids on cytokine production in QuantiFERON-TB Gold In Tube assays.
Tuberculosis (Edinburgh, Scotland) (2015-04-04)
Vanessa Clifford, Christel Zufferey, Susie Germano, Norbert Ryan, David Leslie, Alan Street, Justin Denholm, Marc Tebruegge, Nigel Curtis
ABSTRAKT
The ability to monitor and confirm adequate treatment of latent TB infection (LTBI) would be a major advance. The potential immunomodulatory effects of anti-tuberculous drugs and steroids need to be considered in assessing the utility of cytokine-based assays for this purpose. We determined whether anti-tuberculous antibiotics or dexamethasone affect the production of IFN-γ and other potential cytokine biomarkers (TNF-α, IL-1ra, IL-2, IL-10, IL-13, IP-10, MIP-1β) in the QuantiFERON-TB Gold In-Tube (QFT-IT) assay. Blood from ten adults with LTBI was added to one standard set of QFT-IT tubes and five further sets containing therapeutic concentrations of either isoniazid, rifampicin, isoniazid and rifampicin, ciprofloxacin or dexamethasone. Resulting supernatants were analysed by ELISA (QFT-IT assay IFN-γ) and xMAP-Luminex assays (all cytokines). Anti-tuberculous antibiotics had only a limited effect on categorical QFT-IT assay results and the production of cytokines. In contrast, dexamethasone resulted in a change in categorical results from positive to negative in four of ten patients, and caused a marked reduction in IL-13 and IL-1ra responses. Substantial changes in TB-antigen-induced IFN-γ and other cytokine responses during treatment likely primarily reflect host immunological changes rather than immunomodulatory effects of anti-tuberculous antibiotics. Results from cytokine-based assays in patients on corticosteroids should be interpreted with caution.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%