Merck
Insulin Resistance and Risk of Cardiovascular Disease in Postmenopausal Women: A Cohort Study From the Women's Health Initiative.
Circulation. Cardiovascular quality and outcomes (2015-05-07)
Michelle D Schmiegelow, Haley Hedlin, Marcia L Stefanick, Rachel H Mackey, Matthew Allison, Lisa W Martin, Jennifer G Robinson, Mark A Hlatky
ABSTRAKT
Insulin resistance is associated with diabetes mellitus, but it is uncertain whether it improves cardiovascular disease (CVD) risk prediction beyond traditional cardiovascular risk factors. We identified 15,288 women from the Women's Health Initiative Biomarkers studies with no history of CVD, atrial fibrillation, or diabetes mellitus at baseline (1993-1998). We assessed the prognostic value of adding fasting serum insulin, HOMA-IR (homeostasis model assessment-insulin resistance), serum-triglyceride-to-serum-high-density lipoprotein-cholesterol ratio TG/HDL-C, or impaired fasting glucose (serum glucose ≥110 mg/dL) to traditional risk factors in separate Cox multivariable analyses and assessed risk discrimination and reclassification. The study end point was major CVD events (nonfatal and fatal coronary heart disease and ischemic stroke) within 10 years, which occurred in 894 (5.8%) women. Insulin resistance was associated with CVD risk after adjusting for age and race/ethnicity with hazard ratios (95% confidence interval [CI]) per doubling in insulin of 1.21 (CI, 1.12-1.31), in HOMA-IR of 1.19 (CI, 1.11-1.28), in TG/HDL-C of 1.35 (CI, 1.26-1.45), and for impaired fasting glucose of 1.31 (CI, 1.05-1.64). Although insulin, HOMA-IR, and TG/HDL-C remained associated with increased CVD risk after adjusting for most CVD risk factors, none remained significant after adjusting for HDL-C: hazard ratios for insulin, 1.06 (CI, 0.98-1.16); for HOMA-IR, 1.06 (CI, 0.98-1.15); for TG/HDL-C, 1.11 (CI, 0.99-1.25); and for glucose, 1.20 (CI, 0.96-1.50). Insulin resistance measures did not improve CVD risk discrimination and reclassification. Measures of insulin resistance were no longer associated with CVD risk after adjustment for high-density lipoprotein-cholesterol and did not provide independent prognostic information in postmenopausal women without diabetes mellitus. URL: http://www.clinicaltrial.gov. Unique identifier: NCT00000611.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications