Merck
  • The Endosymbiont Arsenophonus Provides a General Benefit to Soybean Aphid (Hemiptera: Aphididae) Regardless of Host Plant Resistance (Rag).
The Endosymbiont Arsenophonus Provides a General Benefit to Soybean Aphid (Hemiptera: Aphididae) Regardless of Host Plant Resistance (Rag).
Environmental entomology (2015-08-28)
Jason A Wulff, Jennifer A White
ABSTRAKT
Soybean aphid, Aphis glycines Matsumura (Hemiptera: Aphididae), invokes substantial chemical treatment and economic cost in North America. Resistant soybean genotypes hold promise as a low-impact control methodology, but soybean aphid "biotypes" capable of development on resistant soy cast doubt on the durability of soy resistance. We hypothesized that variation in soybean aphid ability to colonize resistant soy is partially attributable to a bacterial symbiont of soybean aphid, Arsenophonus. We used microinjection to manipulate Arsenophonus infection in both virulent and avirulent aphid biotypes, resulting in five pairs of infected versus uninfected isolines. These isolines were subjected to various population growth rate assays on resistant Rag versus susceptible soybean. We found that aphid virulence on Rag soybean was not dependent on Arsenophonus: virulent aphid biotypes performed well on Rag soybean, and avirulent aphid biotypes performed poorly on Rag soybean, regardless of whether Arsenophonus was present or not. However, we did find that Arsenophonus-infected clones on average performed significantly better than their paired uninfected isolines. This pattern was not consistently evident on every date for every clone, either in the population assays nor when we compared lifetime fecundity of individual aphids in a separate experiment. Nevertheless, this overall benefit for infected aphids may be sufficient to explain the high frequency of Arsenophonus infection in soybean aphids.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis