Merck
Antitumor Effect of C21 Steroidal Glycosides on Adenoid Cystic Carcinoma Cell Line SACC83.
Clinical laboratory (2015-12-09)
Qingliang Wang, Jun Cao, Peng Wang, Minghua Ge
ABSTRAKT
To observe the anti-tumor effects and possible mechanism of C21 steroidal glycoside (17-β-Marsdenia tenacissima glycoside B (17-β-tenacigenin B)) and Xiaoaiping injection in the adenoid cystic carcinoma cell line SACC83. We used the salivary adenoid cystic carcinoma cell line SACC83 for the study and used MTT, flow cytometry, and Western-blot (WB) to study the anti-tumor efficacy and mechanism of 17-β-tenacigenin B and Xiaoaiping injection. Three groups were divided as follows: group 1: control group (without drug treatment), group 2: SACC83 treated with 17-β-Marsdenia tenacissima glycoside B, group 3: SACC83 treated with Xiaoaiping injec- tion. The 17-β-tenacigenin B had no significant anti-tumor effect while the Xiaoaiping injection had a dosage-dependent growth inhibition effect on the adenoid cystic carcinoma cell line SACC83. According to WB, after the administration of the 17-p-tenacigenin B, the expression of apoptosis-related protein was not very significant; however, after the administration of the Xiaoaiping injection, the expression of apoptosis inhibition protein bcl-2 decreased while the expression of apoptosis promotion protein bax increased. The 17-β-tenacigenin B had no obvious anti-tumor effects while the Xiaoaiping injection had good anti-tumor effects. Even though the 17-β-tenacigenin B is an important component of the Xiaoaiping injection, in the in vitro experiment, compared with the Xiaoaiping injection, the 17-β-tenacigenin B showed no good anti-tumor effects. However, we need further studies on in vivo experiments of 17-β-tenacigenin B and the anti-tumor effect of other C21 steroidal glycoside monomers in the Xiaoaiping injection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%