Merck
Efficiency of lamellipodia protrusion is determined by the extent of cytosolic actin assembly.
Molecular biology of the cell (2017-03-24)
Georgi Dimchev, Anika Steffen, Frieda Kage, Vanessa Dimchev, Julien Pernier, Marie-France Carlier, Klemens Rottner
ABSTRAKT
Cell migration and cell-cell communication involve the protrusion of actin-rich cell surface projections such as lamellipodia and filopodia. Lamellipodia are networks of actin filaments generated and turned over by filament branching through the Arp2/3 complex. Inhibition of branching is commonly agreed to eliminate formation and maintenance of lamellipodial actin networks, but the regulation of nucleation or elongation of Arp2/3-independent filament populations within the network by, for example, formins or Ena/VASP family members and its influence on the effectiveness of protrusion have been unclear. Here we analyzed the effects of a set of distinct formin fragments and VASP on site-specific, lamellipodial versus cytosolic actin assembly and resulting consequences on protrusion. Surprisingly, expression of formin variants but not VASP reduced lamellipodial protrusion in B16-F1 cells, albeit to variable extents. The rates of actin network polymerization followed a similar trend. Unexpectedly, the degree of inhibition of both parameters depended on the extent of cytosolic but not lamellipodial actin assembly. Indeed, excess cytosolic actin assembly prevented actin monomer from rapid translocation to and efficient incorporation into lamellipodia. Thus, as opposed to sole regulation by actin polymerases operating at their tips, the protrusion efficiency of lamellipodia is determined by a finely tuned balance between lamellipodial and cytosolic actin assembly.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Actin antibody, Mouse monoclonal, clone AC-40, purified from hybridoma cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.