Merck
Aph-1 associates directly with full-length and C-terminal fragments of gamma-secretase substrates.
The Journal of biological chemistry (2010-02-11)
Allen C Chen, Lucie Y Guo, Beth L Ostaszewski, Dennis J Selkoe, Matthew J LaVoie
ABSTRAKT
Gamma-secretase is a ubiquitous, multiprotein enzyme composed of presenilin, nicastrin, Aph-1, and Pen-2. It mediates the intramembrane proteolysis of many type 1 proteins, plays an essential role in numerous signaling pathways, and helps drive the pathogenesis of Alzheimer disease by excising the hydrophobic, aggregation-prone amyloid beta-peptide from the beta-amyloid precursor protein. A central unresolved question is how its many substrates bind and enter the gamma-secretase complex. Here, we provide evidence that both the beta-amyloid precursor protein holoprotein and its C-terminal fragments, the immediate substrates of gamma-secretase, can associate with Aph-1 at overexpressed as well as endogenous protein levels. This association was observed using bi-directional co-immunoprecipitation in multiple systems and detergent conditions, and an beta-amyloid precursor protein-Aph-1 complex was specifically isolated following in situ cross-linking in living cells. In addition, another endogenous canonical gamma-substrate, Jagged, showed association of both its full-length and C-terminal fragment forms with Aph-1. We were also able to demonstrate that this interaction with substrates was conserved across the multiple isoforms of Aph-1 (beta, alphaS, and alphaL), as they were all able to bind beta-amyloid precursor protein with similar affinity. Finally, two highly conserved intramembrane histidines (His-171 and His-197) within Aph-1, which were recently shown to be important for gamma-secretase activity, are required for efficient binding of substrates. Taken together, our data suggest a dominant role for Aph-1 in interacting with gamma-secretase substrates prior to their processing by the proteolytic complex.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Presenilin-1 Antibody, NT, clone hPS1-NT, culture supernatant, clone hPS1-NT, Chemicon®
Informacje o cenach i dostępności nie są obecnie dostępne.