Merck
Role of STAT3 in glucocorticoid-induced expression of the human IL-10 gene.
Molecular immunology (2008-04-12)
Claudia Unterberger, Karl J Staples, Timothy Smallie, Lynn Williams, Brian Foxwell, Annette Schaefer, Bettina Kempkes, T P J Hofer, Max Koeppel, Marion Lohrum, Henk Stunnenberg, Marion Frankenberger, Loems Ziegler-Heitbrock
ABSTRAKT
In the present report we have determined the molecular mechanisms, which govern the expression of the human IL-10 gene when induced by the glucocorticoid Methyl-Prednisolone (MP). Treatment of cells with MP at 10(-6) M will readily induce IL-10 in CD19+ primary B cells and in a human B cell line. Analysis of the IL-10 promoter showed a robust 18-fold induction and demonstrated that a potential GRE motif was not required, while mutation of the -120 STAT-motif strongly reduced MP-induced trans-activation. A strong induction was also seen with a trimeric STAT-motif and over-expression of dominant-negative STAT3 could block MP induction of IL-10 mRNA. Finally, MP treatment induced binding of STAT3 to the promoter as shown by gelshift, supershift and by chromatin-immunoprecipitation. These data show that glucocorticoid-induced expression of the IL-10 gene is mediated by the transcription factor STAT3.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Pepstatin A, microbial, ≥75% (HPLC)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Antipain dihydrochloride from microbial source, protease inhibitor
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder