- Quantification of the steady-state plasma concentrations of clozapine and N-desmethylclozapine in Japanese patients with schizophrenia using a novel HPLC method and the effects of CYPs and ABC transporters polymorphisms.
Quantification of the steady-state plasma concentrations of clozapine and N-desmethylclozapine in Japanese patients with schizophrenia using a novel HPLC method and the effects of CYPs and ABC transporters polymorphisms.
Annals of clinical biochemistry (2016-12-10)
Yumiko Akamine, Yuka Sugawara-Kikuchi, Tsukasa Uno, Tetsuo Shimizu, Masatomo Miura
PMID27932669
ABSTRAKT
Background This study developed a novel high-performance liquid chromatography (HPLC) method for the simultaneous quantification of clozapine and its active metabolite, N-desmethylclozapine, in human plasma and investigated the effects of various factors, including genetic polymorphisms in cytochrome P450 (CYP) 2D6, CYP3A5, ABCB1 and ABCG2, on the steady-state plasma trough concentrations (C
MATERIAŁY
Numer produktu
Marka
Opis produktu
Informacje o cenach i dostępności nie są obecnie dostępne.