Merck
Correlation of lithium bioaccessibility from tea (Camellia sinensis L.) with tea type and consumption habits.
Food chemistry (2017-11-10)
Umran Seven Erdemir, Seref Gucer
ABSTRAKT
In this study, the total contents, leachability into tea infusions, and bioaccessibility of lithium from black, Earl Grey, and green teas were evaluated by inductively coupled plasma mass spectrometry. Leachabilities were evaluated after infusion for 2, 5, or 10min. Bioaccessibility was determined in vitro under simulated stomach and intestinal digestion conditions. Addition of lemon juice, sugar, or milk for consumption, and calcium, tannic acid, and citric acid as additives were evaluated to determine if they affected bioaccessibility of lithium from black tea. The bioaccessible lithium contributed to 0.01%, 0.02%, and 0.03% of the recommended dietary allowances of lithium for black, Earl Grey, and green tea samples, respectively. These contributions may increase up to 4.4 times or decrease up to seven times with certain additives.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Bile extract porcine
Informacje o cenach i dostępności nie są obecnie dostępne.