- Stallion spermatozoa surviving freezing and thawing experience membrane depolarization and increased intracellular Na
Stallion spermatozoa surviving freezing and thawing experience membrane depolarization and increased intracellular Na
Andrology (2017-10-04)
C Ortega Ferrusola, L Anel-López, J M Ortiz-Rodriguez, P Martin Muñoz, M Alvarez, P de Paz, J Masot, E Redondo, C Balao da Silva, J M Morrell, H Rodriguez Martinez, J A Tapia, M C Gil, L Anel, F J Peña
PMID28973824
ABSTRAKT
In order to gain insight of the modifications that freezing and thawing cause to the surviving population of spermatozoa, changes in the potential of the plasma membrane (Em) and intracellular Na