Product Name
Curcumin analytical standard
Product Number
08511
Product Brand
sial
CAS Number
458-37-7
Molecular Weight
368.38
Test
Specification
APPEARANCE (COLOR)Light Yellow to Dark Yellow and Light Orange to Dark Orange
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
IDENTIFICATION (LC-MS)CORRESPONDS TO REQUIREMENTS
EXPIRY PERIOD24 MONTHS