Product Name
DL-Dithiothreitol solution BioUltra, Molecular Biology, ~1 M in H2O
Product Number
43816
Product Brand
sigma
CAS Number
3483-12-3
Molecular Weight
154.25
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
APPEARANCE (TURBIDITY)Clear
TITRATION (MOLARITY)0.9 - 1.1 mol/l
DENSITY D20/41.030 - 1.050
REFRACTIVE INDEX N20/D1.361 - 1.363
EXTRANEOUS ACTIVITIESDNAses, RNAses, Proteases, Phosphatases Not Detectable
SOLUBILITY (METHOD)NEAT
APPEARANCE (SOLUTION)CLEAR COLORLESS
PH (SOLUTION)2.7 - 5.0
RESIDUE (FILTER TEST)NO RESIDUE
CUT-OFF≤ 330.0 nm
RECOMMENDED RETEST PERIOD28 MONTHS