
17α-Hydroxyprogesterone solution

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
Products
Product Category
Brand
Manufacturer
Physical Form
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing