
Maltose monohydrate

Synonym(s):
Maltose monohydrate, 4-O-a-D-Glucopyranosyl-D-glucose
Empirical Formula (Hill Notation):
C12H22O11 · xH2O
CAS Number:
Molecular Weight:
342.30 (anhydrous basis)
Products
Product Category
Brand
Color
Physical Form
Product Line
Purity
Quality Segment
Application
Showing 1-4 of 4