
4-DAMP

Synonym(s):
1,1-Dimethyl-4-diphenylacetoxypiperidinium iodide, 4-DAMP methiodide, 4-Diphenylacetoxy-N-methylpiperidine methiodide, 4-[(Diphenylacetyl)oxy]-1,1-dimethyl-piperidinium iodide
Empirical Formula (Hill Notation):
C21H26INO2
CAS Number:
Molecular Weight:
451.34
Products
Shipping
Product Category
Brand
Color
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing