Skip to Content
MilliporeSigma
Acetone-2-13C 99 atom % 13C

Acetone-2-13C

Acetone-2-13C 99 atom % 13C
Linear Formula:
CH313COCH3
CAS Number:
Molecular Weight:
59.07

Products

Shipping
Product Category
Brand
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing