Skip to Content
MilliporeSigma
BAFF human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

BAFF human

BAFF human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Synonym(s):
TALL-1, THANK, TNFSF13B, TNFSF20

Products

Product Category
Brand
Biological Source
Physical Form
Purity
Showing 1-1 of 1