
FLUVALINATE

Synonym(s):
cyano(3-phenoxyphenyl)methyl (2R)-2-[2-chloro-4-(trifluoromethyl)anilino]-3-methylbutanoate
Linear Formula:
C26H22O3N2Cl1F3
CAS Number:
Molecular Weight:
502.92
Products
Brand
Physical Form
Showing 1-2 of 2
Compare
Product Number
Description
Pricing