
Manool

Synonym(s):
(3R)-5-[(1S,4aS,8aS)-5,5,8a-trimethyl-2-methylenedecahydro-1-naphthalenyl]-3-methyl-1-penten-3-ol
Linear Formula:
C20H34O1
CAS Number:
Molecular Weight:
290.49
Products
Brand
Physical Form
Showing 1-2 of 2
Compare
Product Number
Description
Pricing