Skip to Content
MilliporeSigma
Silver Disk Silver Disk,Formula: AgPercentage Purity: 99.95%Temper: As RolledThickness: 0.3mmDiameter: 20mmCAS Number: 7440-22-4UOM Code: 554-209-47Legacy Code: AG000462Distributor Code: GF55420947

Silver Disk

Silver Disk Silver Disk,Formula: AgPercentage Purity: 99.95%Temper: As RolledThickness: 0.3mmDiameter: 20mmCAS Number: 7440-22-4UOM Code: 554-209-47Legacy Code: AG000462Distributor Code: GF55420947
CAS Number:
Molecular Weight:
0

Products

Brand
Physical Form
Showing 1-20 of 2633
Page 1 of 132
Compare
Product Number
Description
Pricing
Page 1 of 132