Skip to Content
MilliporeSigma
Brassicasterol from semisynthetic

Brassicasterol

Brassicasterol from semisynthetic
Synonym(s):
5,22-Cholestadien-24β-methyl-3β-ol
Empirical Formula (Hill Notation):
C28H46O
CAS Number:
Molecular Weight:
398.66
EC Number:

Products

Product Category
Brand
Biological Source
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing