Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Brix Hydrometer
Synonym(s):
densimeter, gravity meter, salinometer
Products
Product Category
Laboratory Sample Collection
Brand
Sigma-Aldrich
Manufacturer
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z744649
Brix Hydrometer,
Expand
View Pricing
Z744650
Brix Hydrometer,
Expand
View Pricing
Z744647
Brix Hydrometer,
Expand
View Pricing
Z744656
Brix Hydrometer,
Expand
View Pricing
Z744652
Brix Hydrometer,
Expand
View Pricing
Z744653
Brix Hydrometer,
Expand
View Pricing
Z744648
Brix Hydrometer,
Expand
View Pricing
Z744655
Brix Hydrometer,
Expand
View Pricing
Z744651
Brix Hydrometer,
Expand
View Pricing
Z744657
Brix Hydrometer,
Expand
View Pricing
An unknown error has occured.