Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Casein
Products
Product Category
proteins
Brand
Sigma-Aldrich
Physical Form
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.02244
Casein,
Expand
View Pricing
An unknown error has occured.