
Chromium standard solution

Synonym(s):
Chromium Calibration Standard for ICP/AAS
Products
Product Category
Brand
Physical Form
Product Line
Quality Segment
Application
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing