Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Copper - O.F.H.C.
Synonym(s):
Copper, CV000560
Linear Formula:
Cu
CAS Number:
7440-50-8
Molecular Weight:
63.55
Products
Manufacturer
Physical Form
Purity
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
GF95621812
Copper - O.F.H.C.,
foil, 100x100mm, thickness 0.1mm, hard, 99.95+%
Expand
View Pricing
An unknown error has occured.