Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
CTAP
Empirical Formula (Hill Notation):
C
51
H
69
N
13
O
11
S
2
CAS Number:
103429-32-9
Molecular Weight:
1104.30
Products
Product Category
peptides
Brand
Sigma-Aldrich
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C6352
CTAP,
≥95%
Expand
View Pricing
An unknown error has occured.