Skip to Content
MilliporeSigma
Enterodiol ≥95.0% (HPLC)

Enterodiol

Enterodiol ≥95.0% (HPLC)
Synonym(s):
3,3′-[2,3-Bis(hydroxymethyl)butane-1,4-diyl]diphenol
Empirical Formula (Hill Notation):
C18H22O4
CAS Number:
Molecular Weight:
302.36

Products

Product Category
Brand
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing