Skip to Content
MilliporeSigma
Ethidium Bromide OmniPur® Grade, Fluorescent stain

Ethidium Bromide

Ethidium Bromide OmniPur® Grade, Fluorescent stain

Products

Product Category
Brand
Color
Physical Form
Quality Segment
Technique
Showing 1-2 of 2