Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Fluoroshield
™
Products
Product Category
mounting media
Brand
Sigma-Aldrich
Physical Form
Quality Segment
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
F6182
Fluoroshield
™
,
liquid
Expand
View Pricing
F6937
Fluoroshield
™
,
liquid, with 1,4-Diazabicyclo[2.2.2]octane
Expand
View Pricing
An unknown error has occured.