
Fmoc-Lys(Alloc)-OH

Synonym(s):
Nα-Fmoc-Nε-Alloc-L-lysine
Empirical Formula (Hill Notation):
C25H28N2O6
CAS Number:
Molecular Weight:
452.50
Products
Product Category
Brand
Color
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing