Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Formaldehyde-
12
C solution
Synonym(s):
Formalin-
12
C solution
Linear Formula:
H
12
CHO
Molecular Weight:
30.02
Products
Product Category
building blocks
stable isotopes
Brand
Sigma-Aldrich
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
606758
Formaldehyde-
12
C solution,
20% in H
2
O, 99.9 atom %
12
C
Expand
View Pricing
An unknown error has occured.