
FREY Liquid Medium

Synonym(s):
FREY Broth, Mycoplasma Broth, Mycoplasma Liquid Medium
Products
Product Category
Brand
Color
Physical Form
Capacity (mL)
Volume (ml)
Quality Segment
Application
Sterility
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing