Filter Products
Handheld Refractometer, STX-3, 0 to 100 ppt Salinity, ATC
Handheld Refractometer, BX-50, 0 to 50% Brix
Handheld Refractometer, BTX-20, 0 to 20% Brix, ATC
Handheld Refractometer, BX-4, 45 to 82% Brix
Handheld Refractometer, BTX-1, 0 to 32% Brix, ATC
Handheld Refractometer, BX-3, 58 to 92% Brix
Handheld Refractometer, NDX-2, 1.425 to 1.520 nD
Handheld Refractometer, BX-2, 28 to 62% Brix
Handheld Refractometer, CTX-2, Freezing Point °F, ATC
Handheld Refractometer, CTX-1, Freezing Point °C, automatic temperature control
Handheld Refractometer, SGX-1, 0 to 32% Brix / 1.000 to 1.130 SG Beer Wort, ATC
Handheld Refractometer, HMX-1, Honey Moisture, ATC
Handheld Refractometer, SX-2, 0 to 28% NaCl
Handheld Refractometer, URX-60, 0 to 60% Urea
Handheld Refractometer, BX-10, 0 to 10% Brix
Handheld Refractometer, BX-1, 0 to 32% Brix
Handheld Refractometer, BX-20, 0 to 20% Brix
Handheld Refractometer, SX-1, 0 to 10% NaCl