Skip to Content
MilliporeSigma
Vitronectin from human plasma, suitable for cell culture, solution

Vitronectin

Vitronectin from human plasma, suitable for cell culture, solution
Synonym(s):
Cell adhesion protein, ECM protein

Products

Product Category
Brand
Biological Source
Physical Form
Tissue
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing