Skip to Content
MilliporeSigma
Hydroxyproline Assay Kit sufficient for 100 colorimetric tests

Hydroxyproline Assay Kit

Hydroxyproline Assay Kit sufficient for 100 colorimetric tests
Synonym(s):
Hydroxyproline Quantification Kit

Products

Product Category
Brand
Detection Method
Quality Segment
Application
Relevant Disease