Skip to Content
MilliporeSigma
MabSelect™ SuRe™ Cytiva 17-5438-01, pack of 25 mL

MabSelect™ SuRe™

MabSelect™ SuRe™ Cytiva 17-5438-01, pack of 25 mL
Synonym(s):
MabSelect Resin

Products

Product Category
Manufacturer
Physical Form
Technique
Showing 1-2 of 2