
MacConkey agar

Synonym(s):
MacConkey medium
Products
Shipping
Product Category
Brand
Color
Physical Form
Product Line
Diameter (mm)
Volume (ml)
Quality Segment
Application
Sterility
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing