Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Manganese (Mn) standard solution
Synonym(s):
Manganese (Mn) Standard Solution, Manganese nitrate
Products
Product Category
reference materials
Manufacturer
Physical Form
Special Grade
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
NIST3132
Manganese (Mn) standard solution,
NIST
®
SRM
®
3132
Expand
View Pricing
An unknown error has occured.