Skip to Content
MilliporeSigma
Meperidine solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Meperidine solution

Meperidine solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C15H21NO2
CAS Number:
Molecular Weight:
247.33

Products

Product Category
Brand
Manufacturer
Physical Form
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1