Skip to Content
MilliporeSigma
vibrant-m

MILLIPLEX® PLEXpedition Screening Panel

vibrant-m
Synonym(s):
Human screening panel, Luminex human screening panel, Millipore screening kit, Human screening kit, Luminex human screening immunoassay, Millipore screening panel, Multiplex screening kit, Multiplex screening panel

Products

Product Category
Brand
Product Line
Detection Method
Quality Segment
Species Reactivity
Technique