Skip to Content
MilliporeSigma
vibrant-m

N-Nitroso Phenylephrine Solution

vibrant-m
Empirical Formula (Hill Notation):
C9H12N2O3
CAS Number:
Molecular Weight:
196.20

Products

Product Category
Manufacturer
Physical Form
Quality Segment
Application
Special Grade