Skip to Content
MilliporeSigma
Oxalic acid solution for 1000 ml, c(C2H2O4) = 0.05 mol/l (0.1 N) Titrisol®

Oxalic acid solution

Oxalic acid solution for 1000 ml, c(C2H2O4) = 0.05 mol/l (0.1 N) Titrisol®
Synonym(s):
ethanedioic acid solution; dicarboxylic acid solution

Products

Product Category
Brand
Physical Form
Product Line
Quality Segment
Technique