Skip to Content
MilliporeSigma
pH-indicator paper pH 1 - 10 Universal indicator roll L 4.8 m , with color scale, graduations and accuracy accuracy: 1 pH unit, pH range 1-10, MQuant®

pH-indicator paper pH 1 - 10 Universal indicator

pH-indicator paper pH 1 - 10 Universal indicator roll L 4.8 m , with color scale, graduations and accuracy accuracy: 1 pH unit, pH range 1-10, MQuant®
Synonym(s):
pH chemical test paper paper pH 1 - 10 Universal indicator, pH test paper paper pH 1 - 10 Universal indicator

Products

Product Category
Brand
Product Line
Detection Method
Quality Segment
Application
Technique