Skip to Content
MilliporeSigma
Polyethersulfone (PES), granule, 3 mm nominal granule size, weight 200 g, clear amber

Polyethersulfone

Polyethersulfone (PES), granule, 3 mm nominal granule size, weight 200 g, clear amber
Synonym(s):
SU306311

Products

Product Category
Color
Manufacturer
Physical Form
Diameter (mm)
Showing 1-20 of 26
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2