Skip to Content
MilliporeSigma
Polypropylene (PP), film, thickness 0.05 mm, size 600 × 600 mm, condition biaxially oriented

Polypropylene

Polypropylene (PP), film, thickness 0.05 mm, size 600 × 600 mm, condition biaxially oriented
Synonym(s):
PP301350

Products

Shipping
Product Category
Color
Manufacturer
Physical Form
Diameter (mm)
Special Grade
Showing 1-20 of 191
Page 1 of 10
Compare
Product Number
Description
Pricing
Page 1 of 10