Skip to Content
MilliporeSigma
(R)-(+)-Limonene solution certified reference material, 2000 μg/mL in methanol, ampule of 1 mL

(R)-(+)-Limonene solution

(R)-(+)-Limonene solution certified reference material, 2000 μg/mL in methanol, ampule of 1 mL
CAS Number:

Products

Product Category
Brand
Physical Form
Product Line
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1